Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212647.2 | NP_997812.1 | 5 - 6 | 827 | 55 | |
GenBank mRNA | BC064648.1 | - | 755 | 55 | ||
BC152200.1 | - | 5 - 6 | 730 | 55 | ||
BC155104.1 | - | 5 - 6 | 738 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |