Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd82b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212663.1 | NP_997828.1 | 6 - 7 | 547 | 62 | |
GenBank mRNA | BC054586.1 | - | 6 - 7 | 547 | 62 | |
BC164453.1 | - | 5 - 6 | 474 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |