Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328059.1 | NP_001314988.1 | 10 - 11 | 1158 | 99 | |
GenBank mRNA | BC057475.1 | - | 10 - 11 | 1129 | 99 | |
BC154320.1 | - | 10 - 11 | 1122 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |