Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328059.1 | NP_001314988.1 | 7 - 8 | 830 | 67 | |
GenBank mRNA | BC057475.1 | - | 7 - 8 | 801 | 67 | |
BC154320.1 | - | 7 - 8 | 794 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |