Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212738.1 | NP_997903.1 | 10 - 11 | 1014 | 66 | |
XM_021470114.1 | XP_021325789.1 | 10 - 11 | 1337 | 66 | ||
GenBank mRNA | AY289606.1 | - | 10 - 11 | 1014 | 66 | |
BC096854.1 | - | 10 - 11 | 1008 | 66 | ||
BC163965.1 | - | 8 - 9 | 921 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |