Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212755.2 | NP_997920.2 | 2 - 3 | 728 | 107 | |
GenBank mRNA | AY262256.1 | - | 2 - 3 | 690 | 107 | |
BC162661.1 | - | 2 - 3 | 629 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |