Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365553.1 | NP_001352482.1 | 10 - 11 | 1258 | 147 | |
NM_212781.2 | NP_997946.1 | 10 - 11 | 1249 | 147 | ||
GenBank mRNA | AY169369.1 | - | 10 - 11 | 1209 | 147 | |
BC081616.1 | - | 10 - 11 | 1258 | 147 | ||
FJ205701.1 | - | 10 - 11 | 1205 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |