Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smtnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212805.2 | NP_997970.1 | 6 - 7 | 1117 | 80 | |
GenBank mRNA | AB110459.1 | - | 6 - 7 | 1100 | 80 | |
BC090828.1 | - | 6 - 7 | 1121 | 80 | ||
BC165356.1 | - | 5 - 6 | 968 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |