Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lnx2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212940.2 | NP_998105.2 | 5 - 6 | 1530 | 69 | |
XM_005173252.3 | XP_005173309.1 | 6 - 7 | 1686 | 69 | ||
XM_005173253.4 | XP_005173310.1 | 5 - 6 | 1544 | 69 | ||
GenBank mRNA | BC139538.1 | - | 5 - 6 | 1530 | 69 | |
FJ156084.1 | - | 5 - 6 | 1517 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |