Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrpkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212991.3 | NP_998156.3 | 8 - 9 | 685 | 71 | |
XM_009301843.3 | XP_009300118.1 | 8 - 9 | 739 | 71 | ||
GenBank mRNA | BC096867.1 | - | 8 - 9 | 665 | 71 | |
BC164732.1 | - | 7 - 8 | 607 | 71 | ||
CT708088.2 | - | 6 - 7 | 523 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |