Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrpkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212991.3 | NP_998156.3 | 9 - 10 | 995 | 78 | |
XM_009301843.3 | XP_009300118.1 | 9 - 10 | 1049 | 78 | ||
GenBank mRNA | BC047844.1 | - | 7 - 8 | 724 | 78 | |
BC096867.1 | - | 9 - 10 | 975 | 78 | ||
BC164732.1 | - | 8 - 9 | 917 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |