Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213015.1 | NP_998180.1 | 4 - 5 | 634 | 72 | |
XM_005157414.4 | XP_005157471.1 | 4 - 5 | 72 | |||
XM_005157415.4 | XP_005157472.1 | 4 - 5 | 72 | |||
GenBank mRNA | BC055511.1 | - | 4 - 5 | 666 | 72 | |
BC153596.1 | - | 4 - 5 | 72 | |||
BC153597.1 | - | 3 - 4 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |