Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213034.3 | NP_998199.1 | 3 - 4 | 393 | 72 | |
GenBank mRNA | BC059677.1 | - | 3 - 4 | 217 | 72 | |
BC125961.1 | - | 2 - 3 | 1277 | 72 | ||
EH440189.1 | - | 3 - 4 | 257 | 72 | ||
EH567726.1 | - | 1 - 2 | 118 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |