Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213079.1 | NP_998244.1 | 7 - 8 | 890 | 65 | |
XM_017358046.2 | XP_017213535.1 | 7 - 8 | 953 | 65 | ||
XM_021479568.1 | XP_021335243.1 | 7 - 8 | 981 | 65 | ||
XM_021479569.1 | XP_021335244.1 | 8 - 9 | 977 | 65 | ||
XM_021479572.1 | XP_021335247.1 | 7 - 8 | 953 | 65 | ||
XM_021479574.1 | XP_021335249.1 | 8 - 9 | 975 | 65 | ||
GenBank mRNA | BC044458.1 | - | 7 - 8 | 890 | 65 | |
BC097078.1 | - | 7 - 8 | 975 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |