Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213155.2 | NP_998320.1 | 3 - 4 | 429 | 80 | |
GenBank mRNA | BC058042.1 | - | 371 | 80 | ||
DT873506.1 | - | 3 - 4 | 433 | 80 |
Gene Symbol: ppp1r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AI722912.1 | - | 1 - 2 | 169 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |