Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpm6aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213200.1 | NP_998365.1 | 6 - 7 | 786 | 92 | |
GenBank mRNA | AY070261.1 | - | 6 - 7 | 786 | 92 | |
BC064281.1 | - | 6 - 7 | 727 | 92 | ||
BC150455.1 | - | 6 - 7 | 810 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |