Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213352.2 | NP_998517.2 | 2 - 3 | 401 | 106 | |
GenBank mRNA | AW184579.1 | - | 1 - 2 | 106 | ||
BC057417.1 | - | 2 - 3 | 106 | |||
BC152216.1 | - | 2 - 3 | 342 | 106 | ||
CB923490.1 | - | 2 - 3 | 301 | 106 | ||
EH449789.1 | - | 2 - 3 | 380 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |