Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacng1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214684.1 | NP_999849.1 | 2 - 3 | 465 | 131 | |
XM_021476780.1 | XP_021332455.1 | 2 - 3 | 520 | 131 | ||
GenBank mRNA | BC044150.1 | - | 2 - 3 | 465 | 131 | |
BC165423.1 | - | 2 - 3 | 343 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |