Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214686.2 | NP_999851.2 | 4 - 5 | 622 | 74 | |
GenBank mRNA | AY180110.2 | - | 4 - 5 | 552 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |