Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prrx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214734.1 | NP_999899.1 | 1 - 2 | 372 | 73 | |
GenBank mRNA | BC053228.1 | - | 1 - 2 | 372 | 73 | |
BC165100.1 | - | 1 - 2 | 280 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |