Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214780.1 | NP_999945.1 | 3 - 4 | 313 | 83 | |
GenBank mRNA | AY391453.1 | - | 3 - 4 | 313 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |