Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sypb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030242.3 | NP_001025413.2 | 8 | 1626 | 136 | |
GenBank mRNA | BC076299.1 | - | 8 | 1447 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |