Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xkrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007406.1 | NP_001007407.1 | 4 | 1954 | 128 | |
GenBank mRNA | BC085430.1 | - | 4 | 1954 | 128 | |
BC139520.1 | - | 3 | 1731 | 128 | ||
BC155740.1 | - | 4 | 1925 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |