Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185087.3 | NP_001172016.3 | 14 - 15 | 2860 | 85 | |
NM_001185089.2 | NP_001172018.2 | 14 - 15 | 2376 | 85 | ||
XM_005607780.3 | XP_005607837.1 | 3066 | 85 | |||
XM_023626768.1 | XP_023482536.1 | 2947 | 85 | |||
XM_023626782.1 | XP_023482550.1 | 2916 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Equus caballus Probe spans exons |