Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110134.1 | NP_001103604.1 | 4 - 5 | 535 | 66 | |
GenBank mRNA | AB364290.1 | - | 4 - 5 | 535 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |