Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130486.2 | NP_001123958.1 | 4 - 5 | 614 | 111 | |
GenBank mRNA | DN507624.1 | - | 4 - 5 | 593 | 111 | |
EF397508.1 | - | 4 - 5 | 448 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Equus caballus Probe spans exons |