Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081803.2 | NP_001075272.2 | 3 - 4 | 417 | 123 | |
XM_023625232.1 | XP_023481000.1 | 428 | 123 | |||
XM_023625233.1 | XP_023481001.1 | 593 | 123 | |||
XM_023625235.1 | XP_023481003.1 | 364 | 123 | |||
GenBank mRNA | DQ318135.1 | - | 3 - 4 | 821 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |