Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081820.1 | NP_001075289.1 | 6 - 7 | 501 | 92 | |
XM_023636843.1 | XP_023492611.1 | 354 | 92 | |||
GenBank mRNA | AB013344.1 | - | 4 - 5 | 308 | 92 | |
AB049188.1 | - | 6 - 7 | 501 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Equus caballus Inventoried Probe spans exons |