Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081854.3 | NP_001075323.1 | 1 - 2 | 93 | 64 | |
NM_001085424.1 | NP_001078893.1 | 1 - 2 | 93 | 64 | ||
XM_014741479.2 | XP_014596965.1 | 839 | 64 | |||
GenBank mRNA | AF249307.1 | - | 1 - 2 | 93 | 64 | |
AF257471.1 | - | 1 - 2 | 93 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |