Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081854.3 | NP_001075323.1 | 3 - 4 | 326 | 100 | |
XM_014741479.2 | XP_014596965.1 | 1072 | 100 | |||
GenBank mRNA | AF249307.1 | - | 3 - 4 | 326 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Equus caballus Probe spans exons |