Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081898.1 | NP_001075367.1 | 1 - 2 | 146 | 101 | |
GenBank mRNA | AY398685.1 | - | 1 - 2 | 146 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |