Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081903.2 | NP_001075372.1 | 2 - 3 | 273 | 124 | |
GenBank mRNA | AY536518.1 | - | 2 - 3 | 274 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |