Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081912.1 | NP_001075381.1 | 2 - 3 | 243 | 64 | |
XM_005599134.3 | XP_005599191.1 | 351 | 64 | |||
GenBank mRNA | DQ167467.1 | - | 2 - 3 | 243 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Equus caballus Probe spans exons |