Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081928.2 | NP_001075397.1 | 1 - 2 | 45 | 79 | |
GenBank mRNA | AF257470.1 | - | 1 - 2 | 59 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |