Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081929.3 | NP_001075398.2 | 4 - 5 | 462 | 69 | |
GenBank mRNA | AF538056.3 | - | 4 - 5 | 462 | 69 | |
BI960890.1 | - | 4 - 5 | 421 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus Probe spans exons |