Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081951.1 | NP_001075420.1 | 5 - 6 | 841 | 76 | |
XM_023655349.1 | XP_023511117.1 | 709 | 76 | |||
GenBank mRNA | U21208.1 | - | 5 - 6 | 841 | 76 |
Gene Symbol: LOC111773844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642359.1 | XP_023498127.1 | 917 | 76 | ||
XM_023642368.1 | XP_023498136.1 | 709 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Equus caballus Probe spans exons |