Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082496.2 | NP_001075965.2 | 4 - 5 | 117 | ||
GenBank mRNA | AF005227.1 | - | 3 - 4 | 117 | ||
AF041975.1 | - | 3 - 4 | 117 | |||
EU438770.1 | - | 3 - 4 | 462 | 117 | ||
KX345376.1 | - | 3 - 4 | 117 | |||
U64794.1 | - | 4 - 5 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |