Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082518.2 | NP_001075987.2 | 6 - 7 | 1053 | 66 | |
GenBank mRNA | AB120394.1 | - | 6 - 7 | 1054 | 66 | |
AB120395.1 | - | 6 - 7 | 1054 | 66 | ||
KC412060.1 | - | 6 - 7 | 1031 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |