Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082519.1 | NP_001075988.1 | 2 - 3 | 189 | 61 | |
GenBank mRNA | AF305617.1 | - | 2 - 3 | 189 | 61 | |
EU000425.1 | - | 2 - 3 | 189 | 61 | ||
EU438769.1 | - | 2 - 3 | 189 | 61 | ||
GU139701.1 | - | 2 - 3 | 189 | 61 | ||
JQ432546.1 | - | 2 - 3 | 189 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |