Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082524.1 | NP_001075993.1 | 1 - 2 | 92 | 121 | |
XM_023649348.1 | XP_023505116.1 | 364 | 121 | |||
GenBank mRNA | AB120412.1 | - | 1 - 2 | 92 | 121 | |
AB120413.1 | - | 1 - 2 | 92 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Equus caballus Inventoried Probe spans exons |