Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILT11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082527.1 | NP_001075996.1 | 1 - 2 | 104 | 88 | |
XM_005596091.3 | XP_005596148.1 | 111 | 88 | |||
XM_005596093.3 | XP_005596150.1 | 111 | 88 | |||
XM_005596094.3 | XP_005596151.1 | 111 | 88 | |||
XM_005596097.3 | XP_005596154.1 | 111 | 88 | |||
GenBank mRNA | AB120409.1 | - | 1 - 2 | 104 | 88 | |
AB120411.1 | - | 1 - 2 | 59 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Equus caballus Probe spans exons |