Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILT11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083593.1 | NP_001077062.1 | 1 - 2 | 45 | 64 | |
XM_023649356.1 | XP_023505124.1 | 323 | 64 | |||
GenBank mRNA | AB120403.1 | - | 1 - 2 | 45 | 64 | |
AB120404.1 | - | 1 - 2 | 45 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Equus caballus Probe spans exons |