Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085434.1 | NP_001078903.1 | 2 - 3 | 395 | 101 | |
GenBank mRNA | X69884.1 | - | 395 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |