Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085434.1 | NP_001078903.1 | 4 - 5 | 757 | 119 | |
GenBank mRNA | JL635190.1 | - | 2 - 3 | 204 | 119 | |
X69884.1 | - | 757 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Equus caballus Probe spans exons |