Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089586.1 | NP_001083055.1 | 2 - 3 | 1680 | 85 | |
GenBank mRNA | DQ497159.1 | - | 2 - 3 | 1680 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Equus caballus Probe spans exons |