Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099762.1 | NP_001093232.1 | 3 - 4 | 641 | 71 | |
GenBank mRNA | AY373259.2 | - | 3 - 4 | 641 | 71 | |
AY499346.1 | - | 3 - 4 | 600 | 71 | ||
JL615906.1 | - | 3 - 4 | 666 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |