Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100179.1 | NP_001093649.1 | 2 - 3 | 406 | 78 | |
GenBank mRNA | EU000412.1 | - | 2 - 3 | 406 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Equus caballus Probe spans exons |