Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101650.1 | NP_001095120.1 | 1 - 2 | 116 | 112 | |
XM_005610265.3 | XP_005610322.1 | 112 | ||||
GenBank mRNA | EU057895.1 | - | 1 - 2 | 116 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Equus caballus Probe spans exons |