Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109680.1 | NP_001103150.1 | 3 - 4 | 432 | 134 | |
XM_023626553.1 | XP_023482321.1 | 1235 | 134 | |||
XM_023626556.1 | XP_023482324.1 | 1956 | 134 | |||
XM_023626563.1 | XP_023482331.1 | 1005 | 134 | |||
GenBank mRNA | EU029660.1 | - | 3 - 4 | 432 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Equus caballus Probe spans exons |