Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109681.1 | NP_001103151.1 | 4 - 5 | 614 | 114 | |
GenBank mRNA | EU029661.1 | - | 4 - 5 | 614 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Equus caballus Probe spans exons |